Ceramic Rods, Round, Solid, AL23, Thermo Scientific
400 mm, 2 mm
Catalog # CAAA32926-QAT
Ceramic Rods, Round, Solid, AL23, Thermo Scientific
Catalog # CAAA32926-QAT
Supplier:  
Ceramic Rods, Round, Solid, AL23, Thermo Scientific
400 mm, 2 mm
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Pure Al2O3 Aluminium oxide, AL23
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.
Solid aluminum rod.
Spécifications
  • Ø:
    2 mm
  • Longueur:
    400 mm