WHEATON® MicroLiter Sample Vial Component Kits, DWK Life Sciences
Vial, 40.00 ml, Amber, Oui, Solid top, White, PTFE/silicone
Catalog # 89498-620
WHEATON® MicroLiter Sample Vial Component Kits, DWK Life Sciences
Catalog # 89498-620
Supplier:  
WHEATON® MicroLiter Sample Vial Component Kits, DWK Life Sciences
WHEATON® MicroLiter Sample Vial Component Kits, DWK Life Sciences
Vial, 40.00 ml, Amber, Oui, Solid top, White, PTFE/silicone
Discontinued
Product Details & Documents
These component kits are shipped with a pack of vials, a pack of caps and a pack of septa.
Available in clear and amber, with polypropylene or phenolic closures and a variety of styles of liners and septa.
Spécifications
  • Avec bouchon:
    Oui
  • Cap Type:
    Solid top
  • Septum:
    PTFE/silicone
  • Capacité:
    40.00 ml
  • Color:
    Amber
  • Désignation:
    Vial
  • Cap Color:
    White