Polymethylpentene Erlenmeyer Flask with Screw Cap | Avantor
Polymethylpentene Erlenmeyer Flask with Screw Cap
250 ml
Catalog # 470211-824
Fournisseur #:  4109-0250
Polymethylpentene Erlenmeyer Flask with Screw Cap
Catalog # 470211-824
Polymethylpentene Erlenmeyer Flask with Screw Cap
Polymethylpentene Erlenmeyer Flask with Screw Cap
250 ml
Price
$187.00
Log in pour la disponibilité
Pack of 4 Case of 12
Product Details & Documents
PMP flasks with secure leak-proof seal.
  • Translucent
  • Includes screw cap
  • Resistant to most acids, bases, alcohols, esters, and aldehydes
  • Autoclavable
Spécifications
  • Capacité:
    250 ml