Polymethylpentene Erlenmeyer Flask with Screw Cap
250 ml
Catalog # 470211-824
Supplier: Thermo Fisher Scientific
Fournisseur #: 4109-0250
Polymethylpentene Erlenmeyer Flask with Screw Cap
Catalog # 470211-824
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
PMP flasks with secure leak-proof seal.
- Translucent
- Includes screw cap
- Resistant to most acids, bases, alcohols, esters, and aldehydes
- Autoclavable
Spécifications
- Capacité:250 ml