Dropper Bottles
Dropper Bottles, 8 ml
Catalog # 470211-796
Supplier: Thermo Fisher Scientific
Fournisseur #: 2752-9025
Jump to:
Product Details & Documents
These bottles are made of low-density polyethylene.
- Bottles Dispense about 40 mcL per Drop
- Good Chemical Resistance
- Semi-Transparent Bottles
- Polyethylene Construction
The semi-transparent bottles feature color-coded caps.
They have good chemical resistance and dispense about 40 mcL per drop.
Spécifications
- Capacité:8 ml
- Désignation:Dropper Bottles