PYREX® Watch Glasses
Watch glass, 75 mm
Supplier: Corning
Fournisseur #: 9985-75
Jump to:
Product Details & Documents
PYREX® borosilicate glass.
- High chemical durability
- Prevents contamination of solutions
These watch glasses are molded to the same radius of curvature for convenient stacking. The heavy wall and uniform fire-polished edge provides the mechanical strength. The outside diameter cannot be controlled within close limits and a slight degree of out-of-round can be expected.
They are also used as covers for heated beakers.
Packaging: One package has 12 watch glasses.
Spécifications
- Ø:75 mm
- Désignation:Watch glass
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.