PYREX® Watch Glasses
Watch glass, 125 mm
Catalog # 470211-704
Supplier:   Corning
 :  9985-125
PYREX® Watch Glasses
Catalog # 470211-704
Supplier:  Corning
PYREX® Watch Glasses
PYREX® Watch Glasses
Watch glass, 125 mm
Price
$140.00
Log in pour la disponibilité
Pack of 12 Case of 72
Product Details & Documents
PYREX® borosilicate glass.
  • High chemical durability
  • Prevents contamination of solutions
These watch glasses are molded to the same radius of curvature for convenient stacking. The heavy wall and uniform fire-polished edge provides the mechanical strength. The outside diameter cannot be controlled within close limits and a slight degree of out-of-round can be expected.
They are also used as covers for heated beakers.
 : One package has 12 watch glasses.
Spécifications
  • Ø:
    125 mm
  • Désignation:
    Watch glass