PYREX® Volumetric Pipettes, Color-Coded, Class A | Avantor
PYREX® Volumetric Pipettes, Color-Coded, Class A
Pyrex® transfer pipettes (volumetric pipettes), cap., each, 5 ml
Catalog # 470211-606
   Low Manufacturing Impact
Supplier:   Corning
Fournisseur #:  7100-5
PYREX® Volumetric Pipettes, Color-Coded, Class A
Catalog # 470211-606
Supplier:  Corning
   Low Manufacturing Impact
PYREX® Volumetric Pipettes, Color-Coded, Class A
PYREX® Volumetric Pipettes, Color-Coded, Class A
Pyrex® transfer pipettes (volumetric pipettes), cap., each, 5 ml
Price
$28.00
Log in pour la disponibilité
Each Case of 12
Product Details & Documents
These pipettes are made of high-grade borosilicate glass.
  • Reusable, rugged construction
  • Color-coded
  • Colored enamel graduations and markings
Ruggedly constructed and properly proportioned, pipettes have accuracy tolerances to meet or exceed the strict requirements for Class A Volumetric Ware as indicated by ASTM E969.
Spécifications
  • Capacité:
    5 ml
  • Désignation:
    Pyrex® transfer pipettes (volumetric pipettes), cap., each
  • Cat. No.:
    470211-606
  • Environmental certification:
    I-RECs, ISO 14001 - Monterrey
  • Environmentally Preferable:
    Low Manufacturing Impact