PYREX® Volumetric Flasks
500 ml
Supplier: Corning
Fournisseur #: 5580-500
Jump to:
Product Details & Documents
PYREX® glass, clear, with snap-caps.
- Heavy beads and tubing rings tooled for snap-cap fit
- Graduation line is sharp and permanent
- No snap caps for 1 and 2 ml sizes
Spécifications
- Capacité:500 ml