PYREX® Precision Burets | Avantor
PYREX® Precision Burets
50 ml, 0.10 ml
Catalog # 470211-396
Supplier:   Corning
Fournisseur #:  2122A-50
PYREX® Precision Burets
Catalog # 470211-396
Supplier:  Corning
PYREX® Precision Burets
PYREX® Precision Burets
50 ml, 0.10 ml
Price
318,00 $
Log in pour la disponibilité
Each
Product Details & Documents
Suitable for Use Laboratories Not Requiring the Accuracy of Class A Models.
  • Dispensing Burets
  • Permanent graduation markings
  • Nonstick 2 mm Teflon stopcock ensures a smooth, leak-free operation
  • Dust cover included
Spécifications
  • Intervalle de graduation:
    0.10 ml
  • Capacité:
    50 ml