PYREX® Burets
Geissler Buret Locking Stopcock
  470211-390
 :   Corning
 :  2116-LSO
PYREX® Burets
  470211-390
 :  Corning
PYREX® Burets
PYREX® Burets
Geissler Buret Locking Stopcock
$163.00
Each
Educational grade Pyrex burets.
  • Heavy duty
  • Transparent
Supplied with self-lubricating, leakproof PMP stopcock. This buret is unaffected by most dilute aqueous solutions, except ammonia and HF. It is not recommended for use with ethanol or organic solvents. The clear white markings make dark liquids easier to measure the the Teflon stopcock is less likely to freeze up.
  • Désignation:
    Geissler Buret Locking Stopcock