PYREX® Student Burets with Replaceable Components
Pyrex® Replaceable Buret, 25 ml, 0.10 ml
Supplier: Corning
Fournisseur #: 2116-25
Jump to:
Product Details & Documents
Durable Enough for Repeated Student Use.
- Tooled tops and bottoms
- White enamel graduations
- Teflon® stopcock reduces breakage
The replaceable stopcock assemblies feature a screw thread locking nut and collar and a replaceable tip with ground end. Teflon® stopcock permits replacement of the body or tip. New triangular shipping package reduces breakage in transit. Replacement parts available.
Spécifications
- Plage de graduation:0.10 ml
- Capacité:25 ml
- Désignation:Pyrex® Replaceable Buret