VWR® Compound Trinocular Microscope
Product CA19000-100 is a replacement for 89496-668 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
VWR® Compound Laboratory Microscopes, equipped with Planachromatic Objectives, are ideal for laboratory, routine, and research work.
- Equipped with planachromatic objectives
- Ergonomic, anti-fatigue design
- Siedentopf-type head with a 30° inclination and 360° rotating capabilities
- One-year warranty on electrical parts
- Five-year warranty on mechanical parts and optics
The series offers an ergonomic, anti- fatigue design to enable long hours of comfortable observation.
This VWR® Compound Microscope features a Siedentopf type head with a 30° inclination and 360° rotating capabilities. Standard features include, widefield eyepieces WF10X / 20mm, reversed quintuple nosepiece, plan objectives PL 4X, 10X, 40X S, 100X S-Oil, coaxial coarse and fine focusing adjustment, built in low position coaxial mechanical stage, centerable and focusable 1.25 N.A Abbe condenser, iris diaphragm with filter holder, Koehler illumination halogen 12V / 20W with intensity control, main supply 110V UL approved.
Certifications: C-UL, UL Listed. CSA certified.
Ordering information: Microscope is supplied with all accessories listed above. The microscope is also supplied with a one-year warranty on electrical parts, and a five-year warranty on mechanical parts and optics.
Documents
Certificates
Recherche par numéro de lotSpécifications
- Désignation:VWR® 45 mm Frosted Filter