Corning® Minimum Essential Medium (Mod.) (MEM), Corning
Minimum Essential Medium (MEM), Sodium bicarbonate, NEAA, L-glutamine, sodium pyruvate
  CA11028-536
 :   Corning
 :  10-009-CV
Corning® Minimum Essential Medium (Mod.) (MEM), Corning
  CA11028-536
 :  Corning
 :  
Corning® Minimum Essential Medium (Mod.) (MEM), Corning
Corning® Minimum Essential Medium (Mod.) (MEM), Corning
Minimum Essential Medium (MEM), Sodium bicarbonate, NEAA, L-glutamine, sodium pyruvate
 :  
$241.00
Pack of 6 (500ml)
Minimum Essential Medium (Mod.)(EMEM) based on Basal Medium Eagle (BME) formulation to aid cell propagation.
  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment
It is a common cell culture medium developed from early work using Basal Medium Eagle (BME) with normal mammalian fibroblasts and certain subtypes of HeLa cells. When supplemented with serum, MEM has been used for cultivation of a wide variety of cells grown in monolayers, such as fibroblasts. This formulation contains Earle's salts and with or without L-glutamine.
  • Désignation:
    Minimum Essential Medium (MEM), Sodium bicarbonate, NEAA, L-glutamine, sodium pyruvate