Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
WaterPro® RO Systems, Labconco | Avantor

WaterPro® RO Systems, Labconco

WaterPro RO System, 230 V, 50/60 Hz INDIA
Catalog # 89496-478
Supplier:   Labconco
Fournisseur #:  9075080
WaterPro® RO Systems, Labconco
Catalog # 89496-478
Supplier:  Labconco
WaterPro® RO Systems, Labconco
WaterPro® RO Systems, Labconco

WaterPro RO System, 230 V, 50/60 Hz INDIA

Price
$13,700.00
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents

WaterPro RO Stations feature a prefilter, carbon filter and RO membrane that combine to deliver high-quality reverse osmosis (RO) purified water

WaterPro RO may be connected to laboratory glassware washers, such as SteamScrubber® or FlaskScrubber® Vantage, to provide purified rinse water. Features include a 17L tank, Type 304 stainless steel front panel and control panel.

RO purified water is ideal for laboratory applications such as reagent preparation and glassware rinsing. Water may be dispensed manually from a valve or from an optional dispensing gun (sold separately).

Ordering information: Dispensing gun not included. Prefilter/Carbon Filter Kit 9067201 and Reverse Osmosis Membrane 9078700 are required, but sold separately.

Spécifications
  • Désignation:
    WaterPro RO System, 230 V, 50/60 Hz INDIA
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.