VWR® Bin Liners
Gamma irradiated bin liner, Clear, LDPE, 40×48", 2 mil, Individually bagged and tagged in a double bagged pack
LDPE liners for use in clean and controlled aseptic environments.
- Available non sterile or gamma irradiated
Uses include transport of product and waste disposal.
Documents
- Color:Clear
- Désignation:Gamma irradiated bin liner
- Dimensions:40×48"
- Matière:LDPE
- Épaisseur:2 mil
- Packaging:Individually bagged and tagged in a double bagged pack