KeepIT® Freezer Boxes, Low-Profile, Wheaton
The WHEATON KeepIT® Freezer Boxes provide an ideal method for batching and storing samples
The KeepIT-25 accomodates up to 25 external threaded vials. The KeepIT-81 accomodates up to 81 external threaded and the KeepIT-100 accomodates up to 100 internal threaded vials (1.2 – 2 mL).
Openings for each vial in the bottom of the box facilitate the scanning of WHEATON CryoELITE 2D Data Matrix Bar Code Inserts on CryoELITE vials. Made from Eastman Tritan™ BPA free and shatter resistant resin. Boxes have a standard footrpint that is compatible with liquid nitrogen storage shelves and freezer drawers.
Available in six different colors that match the colors of the WHEATON CryoELITE® Cryogenic Vials, creating alternatives for batching and indentifying groups of samples.
- Nb de places:100
- Color:Blue
- Désignation:KeepIT-100 Freezer Box
- Dimensions:130L×130W×52D mm
- For Vials:Internal 2 ml vials



