Corning® Single-Use Bags for HYPERStack® Vessels, Corning
Media bag, 20 L, ULDPE
Supplier: Corning
Supplier #: 91-200-77
Jump to:
Product Details & Documents
These single-use bags can be connected to tubing via tube welding or by using the pre-assembled multipurpose connectors.
They can be used to add media, trypsin, or quenching substrates to complete cell culture in an entirely closed solution.
Specifications
- Capacité:20 L
- Désignation:Media bag
- Matière:ULDPE