Corning® Single-Use Bags for HYPERStack® Vessels, Corning
Corning® Single-Use Bags for HYPERStack® Vessels, Corning
Media bag, 20 L, ULDPE
Catalog # CA89423-226
Supplier:   Corning
Supplier #:  91-200-77
Corning® Single-Use Bags for HYPERStack® Vessels, Corning
Catalog # CA89423-226
Supplier:  Corning
Corning® Single-Use Bags for HYPERStack® Vessels, Corning
Media bag, 20 L, ULDPE
Price
$400.00
Log in for availability
Each (20l)
Product Details & Documents
These single-use bags can be connected to tubing via tube welding or by using the pre-assembled multipurpose connectors.
They can be used to add media, trypsin, or quenching substrates to complete cell culture in an entirely closed solution.
Specifications
  • Capacité:
    20 L
  • Désignation:
    Media bag
  • Matière:
    ULDPE