Corning® Single-Use Bags for HYPERStack® Vessels, Corning | Avantor
Corning® Single-Use Bags for HYPERStack® Vessels, Corning
Quench bag, 5 L, ULDPE
Catalog # CA89423-224
Supplier:   Corning
Fournisseur #:  91-200-76
Corning® Single-Use Bags for HYPERStack® Vessels, Corning
Catalog # CA89423-224
Supplier:  Corning
Corning® Single-Use Bags for HYPERStack® Vessels, Corning
Quench bag, 5 L, ULDPE
Price
$438.00
Log in pour la disponibilité
Each (5l)
Product Details & Documents
These single-use bags can be connected to tubing via tube welding or by using the pre-assembled multipurpose connectors.
They can be used to add media, trypsin, or quenching substrates to complete cell culture in an entirely closed solution.
Spécifications
  • Capacité:
    5 L
  • Désignation:
    Quench bag
  • Matière:
    ULDPE