Corning® Single-Use Bags for HYPERStack® Vessels, Corning
Trypsin bag, 5 L, ULDPE
Supplier: Corning
Fournisseur #: 91-200-75
Jump to:
Product Details & Documents
These single-use bags can be connected to tubing via tube welding or by using the pre-assembled multipurpose connectors.
They can be used to add media, trypsin, or quenching substrates to complete cell culture in an entirely closed solution.
Spécifications
- Capacité:5 L
- Désignation:Trypsin bag
- Matière:ULDPE