Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Corning® Collection Containers, Corning

Corning® Collection Containers, Corning

5 L, EVA, ⅜ connector, 12" L × ⅜" int.Ø × ⅝" ext.Ø tubing, male MPC connector with ⅜" barb, female MPC cap, ⅜ connector, 12" L × ⅜" int.Ø × ⅝" ext.Ø tubing, female MPC connector with ⅜" barb, male MPC cap, Break-off Injection site
Catalog # CA89423-214
Supplier:   Corning
Supplier #:  91-200-43
Corning® Collection Containers, Corning
Catalog # CA89423-214
Supplier:  Corning
Corning® Collection Containers, Corning

5 L, EVA, ⅜ connector, 12" L × ⅜" int.Ø × ⅝" ext.Ø tubing, male MPC connector with ⅜" barb, female MPC cap, ⅜ connector, 12" L × ⅜" int.Ø × ⅝" ext.Ø tubing, female MPC connector with ⅜" barb, male MPC cap, Break-off Injection site

Price
$254.00
Log in pour la disponibilité
Each (5l)
Product Details & Documents

Corning® collection containers are fabricated to match specific application requirements, and are available in a variety of sizes and connector configurations.

  • Single-use bags
  • Designed to be practical and cost-effective alternatives to rigid-walled containers
  • Ideal for biopharmaceutical applications

Unless otherwise noted in container port descriptions, all port tubing components are chemically resistant, heat sealable flexible tubing.

Spécifications
  • Capacité:
    5 L
  • Matière:
    EVA
  • Port 1:
    ⅜ connector, 12" L × ⅜" int.Ø × ⅝" ext.Ø tubing, male MPC connector with ⅜" barb, female MPC cap
  • Port 2:
    ⅜ connector, 12" L × ⅜" int.Ø × ⅝" ext.Ø tubing, female MPC connector with ⅜" barb, male MPC cap
  • Port 3:
    Break-off Injection site