Corning® Collection Containers, Corning
20 L, EVA, Female MPC connector with ¼ barb + plug, slide clamp, Male MPC connector with ¼ barb + plug, slide clamp, Vinyl cap, swabbable valve, female Luer with barb
Supplier: Corning
Fournisseur #: 91-200-39
Jump to:
Product Details & Documents
Corning® collection containers are fabricated to match specific application requirements, and are available in a variety of sizes and connector configurations.
- Single-use bags
- Designed to be practical and cost-effective alternatives to rigid-walled containers
- Ideal for biopharmaceutical applications
Unless otherwise noted in container port descriptions, all port tubing components are chemically resistant, heat sealable flexible tubing.
Spécifications
- Capacité:20 L
- Matière:EVA
- Port 1:Female MPC connector with ¼ barb + plug, slide clamp
- Port 2:Male MPC connector with ¼ barb + plug, slide clamp
- Port 3:Vinyl cap, swabbable valve, female Luer with barb