Corning® Collection Containers, Corning
10 L, EVA, Female MPC connector with ¼ barb + plug, slide clamp, Male MPC connector with ¼ barb + plug, slide clamp, Vinyl cap, swabbable valve, female Luer with barb
  CA89423-206
 :   Corning
 :  91-200-36
Corning® Collection Containers, Corning
  CA89423-206
 :  Corning
Corning® Collection Containers, Corning
10 L, EVA, Female MPC connector with ¼ barb + plug, slide clamp, Male MPC connector with ¼ barb + plug, slide clamp, Vinyl cap, swabbable valve, female Luer with barb
99,10 $
Each (10l)
Corning® collection containers are fabricated to match specific application requirements, and are available in a variety of sizes and connector configurations.
  • Single-use bags
  • Designed to be practical and cost-effective alternatives to rigid-walled containers
  • Ideal for biopharmaceutical applications
Unless otherwise noted in container port descriptions, all port tubing components are chemically resistant, heat sealable flexible tubing.
  • Capacité:
    10 L
  • Matière:
    EVA
  • Port 1:
    Female MPC connector with ¼ barb + plug, slide clamp
  • Port 2:
    Male MPC connector with ¼ barb + plug, slide clamp
  • Port 3:
    Vinyl cap, swabbable valve, female Luer with barb