Corning® Collection Containers, Corning
10 L, EVA, Female MPC connector with ¼ barb + plug, slide clamp, Male MPC connector with ¼ barb + plug, slide clamp, Vinyl cap, swabbable valve, female Luer with barb
: Corning
: 91-200-36
Corning® collection containers are fabricated to match specific application requirements, and are available in a variety of sizes and connector configurations.
- Single-use bags
- Designed to be practical and cost-effective alternatives to rigid-walled containers
- Ideal for biopharmaceutical applications
Unless otherwise noted in container port descriptions, all port tubing components are chemically resistant, heat sealable flexible tubing.
- Capacité:10 L
- Matière:EVA
- Port 1:Female MPC connector with ¼ barb + plug, slide clamp
- Port 2:Male MPC connector with ¼ barb + plug, slide clamp
- Port 3:Vinyl cap, swabbable valve, female Luer with barb