Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Corning® Collection Containers, Corning

Corning® Collection Containers, Corning

10 L, EVA, Male Luer connector with ⅛ barb + cap, 2' L × ⅛" int.Ø × ¼" ext.Ø tubing, slide clamp, Male MPC connector with ⅜ barb + cap, 6" L × ⅜" int.Ø × ⅝" ext.Ø PCMGS tubing, slide clamp, Female Luer with ³⁄₁₆ barb, swabbable valve, cap
Catalog # CA89423-202
Supplier:   Corning
Supplier #:  91-200-10
Corning® Collection Containers, Corning
Catalog # CA89423-202
Supplier:  Corning
Corning® Collection Containers, Corning

10 L, EVA, Male Luer connector with ⅛ barb + cap, 2' L × ⅛" int.Ø × ¼" ext.Ø tubing, slide clamp, Male MPC connector with ⅜ barb + cap, 6" L × ⅜" int.Ø × ⅝" ext.Ø PCMGS tubing, slide clamp, Female Luer with ³⁄₁₆ barb, swabbable valve, cap

Price
$127.00
Log in pour la disponibilité
Each (10l)
Product Details & Documents

Corning® collection containers are fabricated to match specific application requirements, and are available in a variety of sizes and connector configurations.

  • Single-use bags
  • Designed to be practical and cost-effective alternatives to rigid-walled containers
  • Ideal for biopharmaceutical applications

Unless otherwise noted in container port descriptions, all port tubing components are chemically resistant, heat sealable flexible tubing.

Spécifications
  • Capacité:
    10 L
  • Matière:
    EVA
  • Port 1:
    Male Luer connector with ⅛ barb + cap, 2' L × ⅛" int.Ø × ¼" ext.Ø tubing, slide clamp
  • Port 2:
    Male MPC connector with ⅜ barb + cap, 6" L × ⅜" int.Ø × ⅝" ext.Ø PCMGS tubing, slide clamp
  • Port 3:
    Female Luer with ³⁄₁₆ barb, swabbable valve, cap