Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Corning® Collection Containers, Corning

Corning® Collection Containers, Corning

5 L, EVA, Male Luer connector with ⅛ barb + cap, 2' L × ⅛" int.Ø × ¼" ext.Ø tubing, slide clamp, Male MPC connector with ⅜ barb + cap, 6" L × ⅜" int.Ø × ⅝" ext.Ø PCMGS tubing, slide clamp, Female Luer with ³⁄₁₆ barb, swabbable valve, cap
Catalog # CA89423-200
Supplier:   Corning
Supplier #:  91-200-05
Corning® Collection Containers, Corning
Catalog # CA89423-200
Supplier:  Corning
Corning® Collection Containers, Corning

5 L, EVA, Male Luer connector with ⅛ barb + cap, 2' L × ⅛" int.Ø × ¼" ext.Ø tubing, slide clamp, Male MPC connector with ⅜ barb + cap, 6" L × ⅜" int.Ø × ⅝" ext.Ø PCMGS tubing, slide clamp, Female Luer with ³⁄₁₆ barb, swabbable valve, cap

Price
116,00 $
Log in pour la disponibilité
Each (5l)
Product Details & Documents

Corning® collection containers are fabricated to match specific application requirements, and are available in a variety of sizes and connector configurations.

  • Single-use bags
  • Designed to be practical and cost-effective alternatives to rigid-walled containers
  • Ideal for biopharmaceutical applications

Unless otherwise noted in container port descriptions, all port tubing components are chemically resistant, heat sealable flexible tubing.

Spécifications
  • Capacité:
    5 L
  • Matière:
    EVA
  • Port 1:
    Male Luer connector with ⅛ barb + cap, 2' L × ⅛" int.Ø × ¼" ext.Ø tubing, slide clamp
  • Port 2:
    Male MPC connector with ⅜ barb + cap, 6" L × ⅜" int.Ø × ⅝" ext.Ø PCMGS tubing, slide clamp
  • Port 3:
    Female Luer with ³⁄₁₆ barb, swabbable valve, cap
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.