Corning® Collection Containers, Corning
Corning® Collection Containers, Corning
24 L × ¹/₄ Int.Ø ×³/₈" Ext.Ø PCMGS tubing, Female Luer with ¹/₄" Barb, cap, Slide clamp", 24 L × ¹/₄ Int.Ø ×³/₈" Ext.Ø PCMGS tubing, Male Luer with ¹/₄" Barb, cap, Slide clamp", —
Catalog # CA89423-190
Corning® Collection Containers, Corning
Catalog # CA89423-190
Supplier:  
Corning® Collection Containers, Corning
24 L × ¹/₄ Int.Ø ×³/₈" Ext.Ø PCMGS tubing, Female Luer with ¹/₄" Barb, cap, Slide clamp", 24 L × ¹/₄ Int.Ø ×³/₈" Ext.Ø PCMGS tubing, Male Luer with ¹/₄" Barb, cap, Slide clamp", —
Discontinued
Product Details & Documents
Corning® collection containers are fabricated to match specific application requirements, and are available in a variety of sizes and connector configurations.
  • Single-use bags
  • Designed to be practical and cost-effective alternatives to rigid-walled containers
  • Ideal for biopharmaceutical applications
Unless otherwise noted in container port descriptions, all port tubing components are chemically resistant, heat sealable flexible tubing.
Specifications
  • Port 1:
    24 L × ¹/₄ Int.Ø ×³/₈" Ext.Ø PCMGS tubing, Female Luer with ¹/₄" Barb, cap, Slide clamp"
  • Port 2:
    24 L × ¹/₄ Int.Ø ×³/₈" Ext.Ø PCMGS tubing, Male Luer with ¹/₄" Barb, cap, Slide clamp"
  • Port 3: