SINGLET™ Single-Use pH Buffer, Combination Pack, Hach
Combination Pack, ±0.02, 7.00/10.01
Catalog # CA11028-036
Supplier: Hach
Fournisseur #: 2769820
SINGLET™ Single-Use pH Buffer, Combination Pack, Hach
Catalog # CA11028-036
Supplier: Hach
CAS Number:
Jump to:
Product Details & Documents
Single-use standard solutions eliminate the need to prepare standardized buffer solutions for pH meter calibration
- Ideal for on-site analysis
- Easy to use
- Fresh and reliable
You can have fresh pH buffer solutions without having to purchase and carry deionized or demineralized water. The convenient foil-pouch packaging maintains freshness, and keeps the solutions contaminant-free and ready to travel with you anywhere.
Ordering information: Single-Use pH Buffer Combination Packs are packaged with 20 pouches with 10 of each pH listed per pack.
Spécifications
Spécifications
- Désignation:Combination Pack
- pH:7.00/10.01
- pH Accuracy:±0.02



