Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Battery Electrodes

Battery Electrode, Laboratory, 19 mm x 100 mm
  470206-316
 :  6368000
Battery Electrodes
  470206-316
Battery Electrodes
Battery Electrodes

Battery Electrode, Laboratory, 19 mm x 100 mm

 
$33.25
   
Pack of 12 Case of 2880
 

Shelf Life (months): Indefinite
Storage: Green

 : Please Note: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

 
  • Désignation:
    Battery Electrode
  • Taille:
    19 mm x 100 mm
  • Chemical/Physical Specifications:
    Zinc
  • Grade:
    Laboratory