Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
LeLoop® Inoculating Loops

LeLoop® Inoculating Loops

Non-Calibrated Loop, Large, 5.45 µl (3.57 mm)
Catalog # CA11027-266
Supplier:   Decon Labs
Supplier #:  MP19825
LeLoop® Inoculating Loops
Catalog # CA11027-266
Supplier:  Decon Labs
LeLoop® Inoculating Loops
LeLoop® Inoculating Loops

Non-Calibrated Loop, Large, 5.45 µl (3.57 mm)

Price
$183.00
Log in for availability
Pack of 25
Product Details & Documents

LeLoop®, Decon’s inoculating loops, are made of high-quality, twisted nichrome wire which is designed to cool rapidly after flaming.

  • Small loop: 2.26 mm Int.Ø

  • Medium loop: 2.91 mm IInt.Ø
  • Large loop: 3.57 mm Int.Ø

These loops are more economical than platinum or disposable plastic loops. The loops slide smoothly across the agar surface without tearing or gouging. Decon loops are made to be used with Decon’s brass holders.

Calibrated 1 µl (0.001 ml) and 10 µl (0.01 ml) inoculating loops are individually calibrated in accordance with ASM/APHA via Evans Blue Dye Test which is a CAP approved method. Calibration certificates are sent with calibrated loops or are available from Decon Labs.

Certifications: UL listed.

Specifications
  • Capacité:
    5.45 µl (3.57 mm)
  • Désignation:
    Non-Calibrated Loop, Large
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • 8" Brass Holder
    8" Brass Holder
    CA11027-256
    Log in for availability
    Each - $131.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.