The Pill™ Autoclave Deoderizers
Autoclave Deoderizer
Catalog # CA11027-236
Supplier: Decon Labs
Fournisseur #: 5500
Jump to:
Product Details & Documents
The Pill™ Autoclave deodorizer capsules contain concentrated liquid odor neutralizers which absorb odors instead of just masking them.
Gelatin capsule keeps deodorizing liquid off hands and clothes
- Capsules facilitate accurate unit dosing
- Pleasant fresh scent
One capsule deodorizes 7 cubic feet of autoclave capacity with full load of soiled labware. Gelatin capsules are not suited to small tabletop autoclaves. One capsule is too strong for small tabletop autoclaves. A pungent residual odor of unused deodorizer will be present in units smaller than 5 cubic feet.
Spécifications
- Désignation:Autoclave Deoderizer