The Pill™ Autoclave Deoderizers
Autoclave Deoderizer
Catalog # CA11027-234
 :  5100
The Pill™ Autoclave Deoderizers
Catalog # CA11027-234
Supplier:  
The Pill™ Autoclave Deoderizers
The Pill™ Autoclave Deoderizers
Autoclave Deoderizer
Discontinued
Product Details & Documents
The Pill™ Autoclave deodorizer capsules contain concentrated liquid odor neutralizers which absorb odors instead of just masking them.
  • Gelatin capsule keeps deodorizing liquid off hands and clothes
  • Capsules facilitate accurate unit dosing
  • Pleasant fresh scent
One capsule deodorizes 7 cubic feet of autoclave capacity with full load of soiled labware. Gelatin capsules are not suited to small tabletop autoclaves. One capsule is too strong for small tabletop autoclaves. A pungent residual odor of unused deodorizer will be present in units smaller than 5 cubic feet.
Spécifications
  • Désignation:
    Autoclave Deoderizer