Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Spring Scales | Avantor

Spring Scales

8004-MN - 50 g (0.5 N) graduation intervals, 2000 g (20 N)
Catalog # 89426-004
Supplier:   Ohaus
Fournisseur #:  80000017
Spring Scales
Catalog # 89426-004
Supplier:  Ohaus
Spring Scales

8004-MN - 50 g (0.5 N) graduation intervals, 2000 g (20 N)

Price
$60.71
Log in pour la disponibilité
Each
Product Details & Documents

Ideal for general weighing, force experiments, action-reaction demonstrations.

  • Dial plates display
  • Metric/ounce/newtons

Spring scale is made of ABS housing with metal face plate, pointer, spring and hook. Features large bright colored dial face, easy-to-read black graduations on a frosted aluminum background.

Spécifications
  • Désignation:
    8004-MN - 50 g (0.5 N) graduation intervals
  • Weighing Capacity:
    2000 g (20 N)