Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Spring Scales | Avantor

Spring Scales

8002-MN - 20 g (0.2 N) graduation intervals, 500 g (5 N)
Catalog # 89425-994
Supplier:   Ohaus
Fournisseur #:  80000015
Spring Scales
Catalog # 89425-994
Supplier:  Ohaus
Spring Scales

8002-MN - 20 g (0.2 N) graduation intervals, 500 g (5 N)

Price
$60.71
Log in pour la disponibilité
Each
Product Details & Documents

Ideal for general weighing, force experiments, action-reaction demonstrations.

  • Dial plates display
  • Metric/ounce/newtons

Spring scale is made of ABS housing with metal face plate, pointer, spring and hook. Features large bright colored dial face, easy-to-read black graduations on a frosted aluminum background.

Spécifications
  • Désignation:
    8002-MN - 20 g (0.2 N) graduation intervals
  • Weighing Capacity:
    500 g (5 N)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.