Spring Scales
8001-MN - 10 g (0.1 N) graduation intervals, 250 g (2.5 N)
Catalog # 89425-988
Supplier: Ohaus
Fournisseur #: 80000014
Jump to:
Product Details & Documents
Ideal for general weighing, force experiments, action-reaction demonstrations.
- Dial plates display
- Metric/ounce/newtons
Spring scale is made of ABS housing with metal face plate, pointer, spring and hook. Features large bright colored dial face, easy-to-read black graduations on a frosted aluminum background.
Spécifications
- Désignation:8001-MN - 10 g (0.1 N) graduation intervals
- Weighing Capacity:250 g (2.5 N)
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



