Microscope Slide Staining Dish with Rack
Microscope Slide Staining Dish with Rack, 20 Pcs., 41/8 x 33/8 x 23/4"
  470205-616
 :  900200
Microscope Slide Staining Dish with Rack
  470205-616
Microscope Slide Staining Dish with Rack
Microscope Slide Staining Dish with Rack
Microscope Slide Staining Dish with Rack, 20 Pcs., 41/8 x 33/8 x 23/4"
$251.00
Each Case of 6
Dries Slides Efficiently with Room for Multiple Slides
  • Quality slide making supplies
  • Slide staining tools
This quick-draining glass rack holds ten single slides, 19 slides arranged in a zigzag pattern, or 20 slides back to back. It comes with a glass cover.
  • Désignation:
    Microscope Slide Staining Dish with Rack, 20 Pcs.
  • Dimensions:
    41/8 x 33/8 x 23/4"