Media Bottles
125 ml (4.2 oz.), Media Bottles
470205-570
: 219815
Media bottles are manufactured from borosilicate glass for excellent chemical resistance. A no-drop pour lip facilicates pouring and cleaning. Graduations show approximate volume, while a large writing patch can be used for identifying content.
Each bottle has a black phenolic PTFE rubber-lined cap attached. Caps are autoclavable.
- Capacité:125 ml (4.2 oz.)
- Désignation:Media Bottles