Media Bottles
125 ml (4.2 oz.), Media Bottles
  470205-570
 :  219815
Media Bottles
  470205-570
Media Bottles
Media Bottles
125 ml (4.2 oz.), Media Bottles
753,00 $
Case of 48
Media bottles are manufactured from borosilicate glass for excellent chemical resistance. A no-drop pour lip facilicates pouring and cleaning. Graduations show approximate volume, while a large writing patch can be used for identifying content.
Each bottle has a black phenolic PTFE rubber-lined cap attached. Caps are autoclavable.
  • Capacité:
    125 ml (4.2 oz.)
  • Désignation:
    Media Bottles