Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
PerpetuLab Tables with Book Compartments

PerpetuLab Tables with Book Compartments

Epoxy standing height, Red oak, 60×36×24"
Catalog # 470205-402
Supplier #:  C7606K36N
PerpetuLab Tables with Book Compartments
Catalog # 470205-402
PerpetuLab Tables with Book Compartments
PerpetuLab Tables with Book Compartments

Epoxy standing height, Red oak, 60×36×24"

Price
$2,615.85
Log in for availability
Each
Product Details & Documents

Lab tables with solid oak aprons and 1" epoxy worksurface. Tables feature two compartments.

  • Available in sitting height or standing height
  • 1" epoxy resin work surface
  • Heavy-duty corner braces
  • Rubber leg boots

Tables are shipped knocked down with tops attached and are assembled easily with a wrench.

Certifications: Diversified Woodcrafts is SEFA compliant and MAS Certified Green.

Specifications
  • Base/Leg Material:
    Red oak
  • Désignation:
    Epoxy standing height
  • W×D×H:
    60×36×24"
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.