Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
PerpetuLab Tables with Book Compartments

PerpetuLab Tables with Book Compartments

Epoxy sitting height, Red oak, 48×30×24"
Catalog # 470205-310
Supplier #:  C7106
PerpetuLab Tables with Book Compartments
Catalog # 470205-310
PerpetuLab Tables with Book Compartments
PerpetuLab Tables with Book Compartments

Epoxy sitting height, Red oak, 48×30×24"

Price
$2,045.30
Log in for availability
Each
Product Details & Documents

Lab tables with solid oak aprons and 1" epoxy worksurface. Tables feature two compartments.

  • Available in sitting height or standing height
  • 1" epoxy resin work surface
  • Heavy-duty corner braces
  • Rubber leg boots

Tables are shipped knocked down with tops attached and are assembled easily with a wrench.

Certifications: Diversified Woodcrafts is SEFA compliant and MAS Certified Green.

Specifications
  • Base/Leg Material:
    Red oak
  • Désignation:
    Epoxy sitting height
  • W×D×H:
    48×30×24"
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.