Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
VWR® pH Electrode Arm | Avantor

VWR® pH Electrode Arm

VWR® Heavy Duty Electrode Holder and Stand
Catalog # 470204-504
Supplier:   Antyila Scientific
Fournisseur #:  90027-52
VWR® pH Electrode Arm
Catalog # 470204-504
Supplier:  Antyila Scientific
VWR® pH Electrode Arm
VWR® pH Electrode Arm

VWR® Heavy Duty Electrode Holder and Stand

Price
$635.75
Log in pour la disponibilité
Each
Product Details & Documents

Electrode arm moves in almost any direction, holding up to three electrodes safely and securely in selected positions.

  • Ideal for multiple pH readings
  • Accepts all standard electrodes
  • Pivots 360°, Extends 53 cm (207/8")

The arm articulates at the three points to maintain electrodes in vertical position.

Designed to accept any standard glass, reference, combination, or other electrode — even a temperature compensation probe. Weighted base and spring counter-balance permit smooth, fluid movement with superior stability.

Spécifications
  • Désignation:
    VWR® Heavy Duty Electrode Holder and Stand