Scheibler Desiccators
Scheibler Desiccators, ID, mm: 150
Catalog # 470204-324
Supplier: Variety Glass
Fournisseur #: 682-3
Jump to:
Product Details & Documents
Made of clear glass with thick uniform walls and a heavy, broad flange. Cover with molded knob is ground to fit the flange perfectly. Porcelain plates sold separately.
Spécifications
- Désignation:Scheibler Desiccators, ID, mm: 150