Polymethylpentene Erlenmeyer Flask with Screw Cap | Avantor
Polymethylpentene Erlenmeyer Flask with Screw Cap
1000 ml
Catalog # 470203-530
Fournisseur #:  4109-1000
Polymethylpentene Erlenmeyer Flask with Screw Cap
Catalog # 470203-530
Polymethylpentene Erlenmeyer Flask with Screw Cap
Polymethylpentene Erlenmeyer Flask with Screw Cap
1000 ml
Price
$99.30
Log in pour la disponibilité
Each Case of 6
Product Details & Documents
PMP flasks with secure leak-proof seal.
  • Translucent
  • Includes screw cap
  • Resistant to most acids, bases, alcohols, esters, and aldehydes
  • Autoclavable
Spécifications
  • Capacité:
    1000 ml