Vertical Clean Bench
Product Details & Documents
Provides Professional Safety Standards for Lab Applications
Vertical Clean Bench
Appropriate for applications such as plant tissue culture, media plate preparation, electronics inspection, medical device assembly, and pharmacy drug preparation, this purifier Clean Bench features an aerodynamic air foil and 254 nm UV lamp with a three-way switch providing fluorescent or UV lighting. Bi-fold doors close the sash opening, while UV"opaque and resistant " thick clear tempered safety glass is built as the sash and sides. An ergonomic 10° angled and hinged sash pivots upward, locks to a loading height of 20", and has wiping seals to contain contaminants. Other features include a 99.99% efficient HEPA filter, variable speed built-in blower with motorized impeller and solid state control rated for 10 amps, Minihelic pressure gauge, air diffuser, fluorescent lamp, blower switch, 9.4" high sash opening, and two utility ports with iris openings and plugs. Constructed of UV"resistant", glacier white and gray, dry powder epoxy-coated aluminum and steel, the unit also features a one year warranty. Work surface and stand available seperately.
Epoxy Work Surface
Designed to contain spills, this chemical-resistant work surface is contoured to fit the dimensions of the Vertical Clean Bench enclosure. The black surface does not feature cupsink cutouts. Size: 36"W x 29"D x 1"H.
Base Stand With Casters
This ADA compliant base features telescoping legs and casters for smooth, easy movement between the classroom and lab.
Spécifications
- Désignation:Vertical Clean Bench, Base Stand w/ Casters