Turbidity
Turbidity
LaMotte® Water Test Kit, Turbidity
  470203-128
 :   LaMotte
Supplier #:  7519-01
Turbidity
  470203-128
 :  LaMotte
Turbidity
Turbidity
LaMotte® Water Test Kit, Turbidity
$177.75
Kit of 1
Pre-calibrated test tubes and labware give fast, accurate results in the classroom or in the lab. 
  • Range: Five to 200 JTU
  • Material for 50 Tests
  • Compact, Waterproof Carrying Case Contains Kit
  • Instructions Included
  • Pre-calibrated Test Tubes
A compact, waterproof carrying case contains reagents, labware, and step-by-step instructions.
The tests have a range of 5 to 200 JTU.
  • Désignation:
    LaMotte® Water Test Kit, Turbidity