Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Polypropylene Closures | Avantor

Polypropylene Closures

Polypropylene closures, 13 mm, Natural
Catalog # 470203-046
Fournisseur #:  73660-13
Polypropylene Closures
Catalog # 470203-046
Supplier:  
Polypropylene Closures
Polypropylene Closures

Polypropylene closures, 13 mm, Natural

Discontinued
Product Details & Documents

Polypropylene closures for 13 and 20 mm culture tubes.

  • Autoclavable
  • Color coded
Spécifications
  • Color:
    Natural
  • Désignation:
    Polypropylene closures
  • For Tube O.D.:
    13 mm