PYREX® Erlenmeyer Flask with Screw Cap
1000 ml
Supplier: Corning
Fournisseur #: 4985-1L
Jump to:
Product Details & Documents
Made of PYREX® borosilicate glass, with phenolic screw caps.
- Excellent chemical resistance
- Excellent temperature resistance
- Graduated to show approximate capacity
- With marking spot
Erlenmeyer flasks with phenolic screw caps are ideal for use as a shaker flask or for mixing, media prep and storage needs. Impact resistant caps are equipped with cemented-in, inert white rubber liners which are resistant to steam sterilization. Their uniform wall thickness provides the proper balance between mechanical strength and thermal shock resistance.
Spécifications
- Capacité:1000 ml