Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning

PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning

500 ml, 6
Catalog # 470202-166
Supplier:   Corning
Supplier #:  4060-500
PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning
Catalog # 470202-166
Supplier:  Corning
PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning
PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning

500 ml, 6

Price
$34.10
Log in for availability
Each Case of 144
Product Details & Documents

Manufactured from borosilicate glass.

Tooled mouth ensures uniform stopper fit. Florence, long neck. Normal working temperature is 230 °C (446 °F).

Specifications
  • Capacité:
    500 ml
  • Stopper No.:
    6
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.