Parafilm | Avantor
Parafilm
Parafilm, Roll Size, in. x ft.: 2 x 250
Catalog # 470201-930
Supplier:   Heathrow Scientific
Fournisseur #:  HS234526A
Parafilm
Catalog # 470201-930
Parafilm
Parafilm
Parafilm, Roll Size, in. x ft.: 2 x 250
Price
64,60 $
Log in pour la disponibilité
Each Case of 24
Product Details & Documents
Flexible, self-adhesive, waterproof plastic for closures, mounting and more.
Adheres quickly and firmly to itself, covering culture tube or flask mouths and sealing petri dish edges. Excellent resistance to air, other gases, alcohol, common acids, and moderately concentrated alkalies.
Spécifications
  • Désignation:
    Parafilm, Roll Size, in. x ft.: 2 x 250