Disposable Polypropylene Snap Cap Test Tubes
Disposable Polypropylene Snap Cap Test Tubes
Disposable Polypropylene Snap Cap Test Tubes, 12 x 75 mm, 5 mL Cap
Catalog # 470201-700
Supplier:   Corning
Supplier #:  352063
Disposable Polypropylene Snap Cap Test Tubes
Catalog # 470201-700
Supplier:  Corning
Disposable Polypropylene Snap Cap Test Tubes
Disposable Polypropylene Snap Cap Test Tubes
Disposable Polypropylene Snap Cap Test Tubes, 12 x 75 mm, 5 mL Cap
Price
$16.70
Log in pour la disponibilité
Pack of 25 Case of 500
Product Details & Documents
Ensured sterility with the convenience of disposability makes these tubes ideal for all types of laboratory procedures.
Spécifications
  • Désignation:
    Disposable Polypropylene Snap Cap Test Tubes, 12 x 75 mm, 5 mL Cap