Flask Brushes
White, Plastic, 17"
Catalog # 470201-504
Supplier: Allied K & R
Fournisseur #: D132
Jump to:
Product Details & Documents
White bristle and plastic handle.
- Multiple sizes available
Recommended for volumetric and boiling flasks because of the unusally flexible, curved steel spring that enables the brush to reach all parts of the flask.
Spécifications
- Bristle Color:White
- Handle Material:Plastic
- Longueur totale:17"