Bare Copper Wire, 113 g Spool
Bare Copper Wire — 113g Spool
Catalog # 470201-430
Fournisseur #: B20
Jump to:
Discontinued
Product Details & Documents
A bare copper wire of the highest quality. Available in several standard AWG sizes.
- 113 g spool
Spécifications
- Désignation:Bare Copper Wire — 113g Spool



