Introductory Plant Microscope Slides
Le produit 470177-830 est similaire à 470201-416 mais peut avoir un fabricant et/ou un emballage différents.
Product Details & Documents
Students explore plant structures and develop microscopy skills with these slides.
- Rigorous Quality Control Standards
- Clearly Distinguishable Identifying Features
- Economically Priced Slides
- Slides Help Students Develop Microscopy Skills
When first learning about microscopy and biology, students need to practice observing and classifying basic structures. These economy slides are designed specifically for introducing life science concepts. The mounted specimens can be clearly viewed in detail to help students gain a set of foundational knowledge and skills they can apply to further scientific study.
The slides are economically priced for bulk or repeat purchases in case of breakage.
Spécifications
- Type:Plant
- Désignation:Leaf epidermis (shows stomates) slide
- Stain:—
- Orientation:Section